Ethyl-2-naphthoat

Lieferant: FLUOROCHEM

018687-10G
FLUO018687-10GEA 438 EUR
FLUO018687-10G
Ethyl-2-naphthoat
Ethyl-2-naphthoat
Formel: C₁₃H₁₂O₂
Molecular Weight: 200,23 g/mol
MDL: MFCD00016811
CAS-Nummer: 3007-91-8
EINECS: 221-119-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR