Ethyl-3-nitrobenzoat
Lieferant: FLUOROCHEM
Synonyme:
3-Nitrobenzoesäureethylester
018688-10G
FLUO018688-10GEA
65.3
EUR
FLUO018688-10G
Ethyl-3-nitrobenzoat
Ethyl-3-nitrobenzoat
Formel:
C₉H₉NO₄ Molecular Weight: 195,17 g/mol |
MDL:
MFCD00014702 CAS-Nummer: 618-98-4 EINECS: 210-574-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...