(4-Nitrophenyl)essigsäureethylester

Lieferant: FLUOROCHEM

Synonyme: Ethyl-(4-nitrophenyl)acetat

018689-10G
FLUO018689-10GEA 156 EUR
FLUO018689-10G
(4-Nitrophenyl)essigsäureethylester
(4-Nitrophenyl)essigsäureethylester
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209,2 g/mol
MDL: MFCD00017046
CAS-Nummer: 5445-26-1
EINECS: 226-646-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR