Ethylpropionylacetat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 3-oxopentanoate, Ethyl 3-oxovalerate
018694-10G
FLUO018694-10GEA
82.1
EUR
FLUO018694-10G
Ethylpropionylacetat
Ethylpropionylacetat
Formel:
C₇H₁₂O₃ Molecular Weight: 144,17 g/mol |
MDL:
MFCD00009317 CAS-Nummer: 4949-44-4 EINECS: 225-593-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...