Pentifyllin

Lieferant: FLUOROCHEM

Synonyme: 1-Hexyl-3,7-dimethyl-3,7-dihydro-2H-purin-2,6-dion, 2,6-Dihydroxy-3,7-dimethyl-1-hexylpurin, 1-Hexyl-3,7-dimethyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dimethyl-1-hexyl-1H,3H-purin-2,6-dion, 3,7-Dimethyl-1-hexylxanthin, 1-Hexyl-3,7-dimethylxanthin, 3,7-Dimethyl-1-hexyl-3,7-dihydro-1H-purin-2,6-dion, 1-Hexyl-3,7-dimethyl-1H,3H-purin-2,6-dion, 1-Hexyltheobromin, 3,7-Dimethyl-1-hexyl-3,7-dihydro-2H-purin-2,6-dion

018709-1G
FLUO018709-1GEA 82.1 EUR
FLUO018709-1G
Pentifyllin
Pentifyllin
Formel: C₁₃H₂₀N₄O₂
Molecular Weight: 264.33 g/mol
MDL: MFCD00041424
CAS-Nummer: 1028-33-7
EINECS: 213-842-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR