4-Ethyl-3-thiosemicarbazid

Lieferant: FLUOROCHEM

Synonyme: 4-Ethyl-3-thiosemicarbazid

018710-1G
FLUO018710-1GEA 45.9 EUR
FLUO018710-1G
4-Ethyl-3-thiosemicarbazid
4-Ethyl-3-thiosemicarbazid
Formel: C₃H₉N₃S
Molecular Weight: 119,19 g/mol
Siedepunkt: 221 °C (1013 hPa)
Schmelzpunkt: 84 °C
MDL: MFCD00007618
CAS-Nummer: 13431-34-0
EINECS: 236-553-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR