1-Hexyl-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: N-Hexylthioharnstoff

018711-1G
FLUO018711-1GEA 73.1 EUR
FLUO018711-1G
1-Hexyl-2-thioharnstoff
1-Hexyl-2-thioharnstoff
Formel: C₇H₁₆N₂S
Molecular Weight: 160.28 g/mol
Siedepunkt: 259
Schmelzpunkt: 83
MDL: MFCD00041198
CAS-Nummer: 21071-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR