2-Hydroxyethyl-4-nitrophenylsulfid

Lieferant: FLUOROCHEM

018716-5G
FLUO018716-5GEA 245 EUR
FLUO018716-5G
2-Hydroxyethyl-4-nitrophenylsulfid
2-Hydroxyethyl-4-nitrophenylsulfid
Formel: C₈H₉NO₃S
MDL: MFCD00060548
CAS-Nummer: 13287-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR