2-Hydroxy-5-methylisophthalaldehyd

Lieferant: FLUOROCHEM

Synonyme: 2-Hydroxy-5-methylbenzene-1,3-dicarbaldehyde

018720-1G
FLUO018720-1GEA 162 EUR
FLUO018720-1G
2-Hydroxy-5-methylisophthalaldehyd
2-Hydroxy-5-methylisophthalaldehyd
Formel: C₉H₈O₃
Molecular Weight: 164,1600 g/mol
MDL: MFCD00016596
CAS-Nummer: 7310-95-4
EINECS: 230-768-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR