(4-Hydroxy-3-nitrophenyl)essigsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-(4-Hydroxy-3-nitrophenyl)acetic acid
018724-1G
FLUO018724-1GEA
45.8
EUR
FLUO018724-1G
(4-Hydroxy-3-nitrophenyl)essigsäure
(4-Hydroxy-3-nitrophenyl)essigsäure
Formel:
(C₆H₃(OH)(NO₂))CH₂COOH Molecular Weight: 197,15 g/mol Schmelzpunkt: 149…150 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00007122 CAS-Nummer: 10463-20-4 EINECS: 233-948-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...