trans-4-Hydroxy-β-nitrostyrol

Lieferant: FLUOROCHEM

Synonyme: (E)-4-(2-Nitrovinyl)phenol

018725-2G
FLUO018725-2GEA 0 EUR
FLUO018725-2G
trans-4-Hydroxy-β-nitrostyrol
trans-4-Hydroxy-β-nitrostyrol
Formel: C₈H₇NO₃
Molecular Weight: 165,1461 g/mol
MDL: MFCD00060551
CAS-Nummer: 22568-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR