(4-Hydroxyphenyl)acetonitril
Lieferant: FLUOROCHEM
Synonyme:
4-Hydroxybenzylcyanid
018726-1G
FLUO018726-1GEA
0
EUR
FLUO018726-1G
(4-Hydroxyphenyl)acetonitril
(4-Hydroxyphenyl)acetonitril
Formel:
C₈H₇NO Siedepunkt: 330 °C (1013 hPa) Schmelzpunkt: 68…71 °C |
MDL:
MFCD00002383 CAS-Nummer: 14191-95-8 EINECS: 238-046-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...