3-(2-Hydroxyphenyl)propionsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(2-Hydroxyphenyl)propansäure
018727-5G
FLUO018727-5GEA
48.8
EUR
FLUO018727-5G
3-(2-Hydroxyphenyl)propionsäure
3-(2-Hydroxyphenyl)propionsäure
Formel:
C₉H₁₀O₃ |
MDL:
MFCD00014026 CAS-Nummer: 495-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...