3-(3-Hydroxyphenyl)propionsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(3-Hydroxyphenyl)propanoic acid

018728-5G
FLUO018728-5GEA 130 EUR
FLUO018728-5G
3-(3-Hydroxyphenyl)propionsäure
3-(3-Hydroxyphenyl)propionsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00016554
CAS-Nummer: 621-54-5
EINECS: 210-692-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR