3-(4-Hydroxyphenyl)propiononitril
Lieferant: FLUOROCHEM
Synonyme:
3-(4-Hydroxyphenyl)propionitrile
018729-10G
FLUO018729-10GEA
303
EUR
FLUO018729-10G
3-(4-Hydroxyphenyl)propiononitril
3-(4-Hydroxyphenyl)propiononitril
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol |
MDL:
MFCD00016492 CAS-Nummer: 17362-17-3 EINECS: 241-393-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...