1-(3-Hydroxyphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM
Achtung

018732-10G
FLUO018732-10GEA 410 EUR
FLUO018732-10G
1-(3-Hydroxyphenyl)-2-thioharnstoff
1-(3-Hydroxyphenyl)-2-thioharnstoff
Formel: (C₆H₄(OH))NC(S)NH₂
Molecular Weight: 168,22 g/mol
MDL: MFCD00022167
CAS-Nummer: 3394-05-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR