1-(4-Hydroxyphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
N-(4-Hydroxyphenyl)thiourea, 4-Hydroxyphenylthiourea
018733-10G
FLUO018733-10GEA
371
EUR
FLUO018733-10G
1-(4-Hydroxyphenyl)-2-thioharnstoff
1-(4-Hydroxyphenyl)-2-thioharnstoff
Formel:
(C₆H₄(OH))NC(S)NH₂ Molecular Weight: 168,22 g/mol |
MDL:
MFCD00041186 CAS-Nummer: 1520-27-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...