4-Iodbenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 4-Iodbenzohydrazid, 4-Iodbenzol-1-carbohydrazid, 4-Iodbenzoesäurehydrazid

018736-5G
FLUO018736-5GEA 42.9 EUR
FLUO018736-5G
4-Iodbenzhydrazid
4-Iodbenzhydrazid
Formel: C₇H₇IN₂O
MDL: MFCD00270107
CAS-Nummer: 39115-95-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR