3-Iod-o-toluylsäure

Lieferant: FLUOROCHEM

018744-10G
FLUO018744-10GEA 180 EUR
FLUO018744-10G
3-Iod-o-toluylsäure
3-Iod-o-toluylsäure
Formel: C₈H₇IO₂
Molecular Weight: 262,05 g/mol
MDL: MFCD00270112
CAS-Nummer: 133232-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR