(Iodmethyl)trimethylammoniumiodid

Lieferant: FLUOROCHEM

018747-2G
FLUO018747-2GEA 0 EUR
FLUO018747-2G
(Iodmethyl)trimethylammoniumiodid
(Iodomethyl)trimethylammonium iodide
Formel: C₄H₁₁I₂N
MDL: MFCD00031647
CAS-Nummer: 39741-91-8
EINECS: 254-611-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR