1-Pentyl-4-iodbenzol
Lieferant: FLUOROCHEM
Synonyme:
1-(4'-Iodphenyl)pentan, 1-(4-Iodphenyl)pentan, 1-Iod-4-pentylbenzol
018749-5G
FLUO018749-5GEA
251
EUR
FLUO018749-5G
1-Pentyl-4-iodbenzol
1-Pentyl-4-iodbenzol
Formel:
C₁₁H₁₅I Molecular Weight: 274,14 g/mol |
MDL:
MFCD00079767 CAS-Nummer: 85017-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...