1-(2-Isopropylphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
Synonyme:
1-(2-Isopropylphenyl)thiourea
018756-5G
FLUO018756-5GEA
171
EUR
FLUO018756-5G
1-(2-Isopropylphenyl)-2-thioharnstoff
1-(2-Isopropylphenyl)-2-thioharnstoff
Formel:
C₁₀H₁₄NS Molecular Weight: 194.2966 g/mol |
MDL:
MFCD00060436 CAS-Nummer: 25343-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...