1-(4-Methylphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

018757-1G
FLUO018757-1GEA 72.9 EUR
FLUO018757-1G
1-(4-Methylphenyl)-2-thioharnstoff
1-(4-Methylphenyl)-2-thioharnstoff
Formel: C₁₀H₁₄N₂S
MDL: MFCD00060459
CAS-Nummer: 65259-91-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR