o-Anisohydrazid

Lieferant: FLUOROCHEM

018769-1G
FLUO018769-1GEA 0 EUR
FLUO018769-1G
o-Anisohydrazid
o-Anisohydrazid
Formel: C₈H₁₀N₂O₂
MDL: MFCD00014755
CAS-Nummer: 7466-54-8
EINECS: 231-260-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR