m-Anisohydrazid

Lieferant: FLUOROCHEM

Synonyme: 3-Methoxybenzoesäurehydrazid, 3-Methoxybenzhydrazid, 3-Methoxybenzol-1-carbohydrazid, 3-Anisohydrazid, 3-Methoxybenzohydrazid

018770-10G
FLUO018770-10GEA 49.4 EUR
FLUO018770-10G
m-Anisohydrazid
m-Anisohydrazid
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166.18 g/mol
MDL: MFCD00007601
CAS-Nummer: 5785-06-8
EINECS: 227-310-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR