5-Methoxybenzofuran

Lieferant: FLUOROCHEM

018772-1G
FLUO018772-1GEA 233 EUR
FLUO018772-1G
5-Methoxybenzofuran
5-Methoxybenzofuran
Formel: C₉H₈O₂
Molecular Weight: 148,16 g/mol
MDL: MFCD00079770
CAS-Nummer: 13391-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR