7-Methoxybenzofuran

Lieferant: FLUOROCHEM

018773-1G
FLUO018773-1GEA 288 EUR
FLUO018773-1G
7-Methoxybenzofuran
7-Methoxybenzofuran
Formel: C₉H₈O₂
Molecular Weight: 148,16 g/mol
MDL: MFCD01310825
CAS-Nummer: 7168-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR