5-Methoxy-2-benzofurancarbonsäure

Lieferant: FLUOROCHEM

018774-1G
FLUO018774-1GEA 101 EUR
FLUO018774-1G
5-Methoxy-2-benzofurancarbonsäure
5-Methoxybenzofuran-2-carboxylic acid
Formel: C₁₀H₈O₄
Molecular Weight: 192,1700 g/mol
MDL: MFCD00079771
CAS-Nummer: 10242-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR