7-Methoxy-2-benzofurancarbonsäure

Lieferant: FLUOROCHEM

018775-1G
FLUO018775-1GEA 67.1 EUR
FLUO018775-1G
7-Methoxy-2-benzofurancarbonsäure
7-Methoxybenzofuran-2-carboxylic acid
Formel: C₁₀H₈O₄
Molecular Weight: 192,17 g/mol
MDL: MFCD00060516
CAS-Nummer: 4790-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR