Ethyl-5-methoxy-2-benzofurancarboxylat

Lieferant: FLUOROCHEM

018776-1G
FLUO018776-1GEA 155 EUR
FLUO018776-1G
Ethyl-5-methoxy-2-benzofurancarboxylat
Ethyl-5-methoxy-2-benzofurancarboxylat
Formel: C₁₂H₁₂O₄
MDL: MFCD00079772
CAS-Nummer: 50551-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR