2-Methoxybenzophenon
Lieferant: FLUOROCHEM
018778-10G
FLUO018778-10GEA
251
EUR
FLUO018778-10G
2-Methoxybenzophenon
2-Methoxybenzophenon
Formel:
C₁₄H₁₂O₂ |
MDL:
MFCD00017163 CAS-Nummer: 2553-04-0 EINECS: 219-857-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...