4-Methoxybiphenyl

Lieferant: FLUOROCHEM

Synonyme: Biphenyl-4-yl methyl ether, 4-Phenylanisole

018785-10G
FLUO018785-10GEA 0 EUR
FLUO018785-10G
4-Methoxybiphenyl
4-Methoxybiphenyl
Formel: C₁₃H₁₂O
Molecular Weight: 184,24 g/mol
MDL: MFCD00014897
CAS-Nummer: 613-37-6
EINECS: 210-339-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR