1-(2-Methoxyethyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Methoxyethyl)thiourea

018788-5G
FLUO018788-5GEA 299 EUR
FLUO018788-5G
1-(2-Methoxyethyl)-2-thioharnstoff
1-(2-Methoxyethyl)-2-thioharnstoff
Formel: C₄H₁₀N₂OS
Molecular Weight: 134.2000 g/mol
MDL: MFCD00060469
CAS-Nummer: 102353-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR