4-Methoxy-3-methylphenylessigsäure

Lieferant: FLUOROCHEM

018791-1G
FLUO018791-1GEA 39.9 EUR
FLUO018791-1G
4-Methoxy-3-methylphenylessigsäure
4-Methoxy-3-methylphenylessigsäure
Formel: C₁₀H₁₂O₃
MDL: MFCD00041034
CAS-Nummer: 4513-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR