trans-2-Methoxy-β-nitrostyrol
Lieferant: FLUOROCHEM
018794-10G
FLUO018794-10GEA
200
EUR
FLUO018794-10G
trans-2-Methoxy-β-nitrostyrol
trans-2-Methoxy-β-nitrostyrol
Formel:
C₉H₉NO₃ |
MDL:
MFCD00070647 CAS-Nummer: 3316-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...