(3-Methoxyphenyl)aceton

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Methoxyphenyl)propan-2-one

018796-1G
FLUO018796-1GEA 75.9 EUR
FLUO018796-1G
(3-Methoxyphenyl)aceton
(3-Methoxyphenyl)aceton
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2 g/mol
MDL: MFCD00008771
CAS-Nummer: 3027-13-2
EINECS: 221-191-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR