(3-Methoxyphenyl)acetonitril

Lieferant: FLUOROCHEM

Synonyme: 3-Methoxybenzylcyanid

018797-10G
FLUO018797-10GEA 39.8 EUR
FLUO018797-10G
(3-Methoxyphenyl)acetonitril
(3-Methoxyphenyl)acetonitril
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
Siedepunkt: 164 °C (1013 hPa)
Dichte: 1,05 g/cm³ (20 °C)
MDL: MFCD00001910
CAS-Nummer: 19924-43-7
EINECS: 243-428-5
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR