2-(2-Methoxyphenyl)ethanol

Lieferant: FLUOROCHEM

018798-10G
FLUO018798-10GEA 321 EUR
FLUO018798-10G
2-(2-Methoxyphenyl)ethanol
2-(2-Methoxyphenyl)ethanol
Formel: C₉H₁₂O₂
MDL: MFCD00004512
CAS-Nummer: 7417-18-7
EINECS: 231-029-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR