1-(4-Methoxyphenyl)imidazol

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Methoxyphenyl)-1H-imidazol

018799-1G
FLUO018799-1GEA 128 EUR
FLUO018799-1G
1-(4-Methoxyphenyl)imidazol
1-(4-Methoxyphenyl)imidazole
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174.2 g/mol
MDL: MFCD00060492
CAS-Nummer: 10040-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR