3-(2-Methoxyphenyl)propionsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(2-Methoxyphenyl)propansäure
018801-5G
FLUO018801-5GEA
45.5
EUR
FLUO018801-5G
3-(2-Methoxyphenyl)propionsäure
3-(2-Methoxyphenyl)propionsäure
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2 g/mol Siedepunkt: 302 °C (1013 hPa) Schmelzpunkt: 92 °C |
MDL:
MFCD00002772 CAS-Nummer: 6342-77-4 EINECS: 228-738-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...