3-(3-Methoxyphenyl)propionsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(3-Methoxyphenyl)propansäure

018802-10G
FLUO018802-10GEA 140 EUR
FLUO018802-10G
3-(3-Methoxyphenyl)propionsäure
3-(3-Methoxyphenyl)propionsäure
Formel: C₁₀H₁₂O₃
MDL: MFCD00014027
CAS-Nummer: 10516-71-9
EINECS: 234-049-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR