1-(3-Methoxyphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM
Achtung

Synonyme: N-(3-Methoxyphenyl)thioharnstoff, 3-Methoxyphenylthioharnstoff

018809-10G
FLUO018809-10GEA 288 EUR
FLUO018809-10G
1-(3-Methoxyphenyl)-2-thioharnstoff
1-(3-Methoxyphenyl)-2-thioharnstoff
Formel: (C₆H₄(OCH₃))NC(S)NH₂
Molecular Weight: 182,25 g/mol
MDL: MFCD00060446
CAS-Nummer: 37014-08-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR