1-Methylbenzotriazol

Lieferant: FLUOROCHEM

Synonyme: 1-Methyl-1H-benzotriazole

018820-10G
FLUO018820-10GEA 285 EUR
FLUO018820-10G
1-Methylbenzotriazol
1-Methylbenzotriazole
Formel: C₇H₇N₃
MDL: MFCD00014572
CAS-Nummer: 13351-73-0
EINECS: 236-401-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR