Methyl-4-benzyloxybenzoat

Lieferant: FLUOROCHEM

018821-10G
FLUO018821-10GEA 225 EUR
FLUO018821-10G
Methyl-4-benzyloxybenzoat
Methyl-4-benzyloxybenzoat
Formel: C₁₅H₁₄O₃
Molecular Weight: 242,27 g/mol
MDL: MFCD00017613
CAS-Nummer: 32122-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR