Methyl-4-phenylbenzoat

Lieferant: FLUOROCHEM

Synonyme: p-Phenylbenzoic acid-OMe, Methyl biphenyl-4-carboxylate, Methyl [1,1'-biphenyl]-4-carboxylate

018823-5G
FLUO018823-5GEA 0 EUR
FLUO018823-5G
Methyl-4-phenylbenzoat
Methyl-4-phenylbenzoat
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
MDL: MFCD00017200
CAS-Nummer: 720-75-2
EINECS: 211-954-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR