Methyl-2-chlorbenzoat

Lieferant: FLUOROCHEM

Synonyme: 2-Chlorobenzoic acid emthyl ester

018825-25G
FLUO018825-25GEA 46.2 EUR
FLUO018825-25G
Methyl-2-chlorbenzoat
Methyl-2-chlorbenzoat
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
Dichte: 1,247 g/cm³ (20 °C)
MDL: MFCD00016337
CAS-Nummer: 610-96-8
EINECS: 210-242-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR