Methyl-2-chlor-4-nitrobenzoat
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-4-nitrobenzoesäuremethylester
018828-10G
FLUO018828-10GEA
139
EUR
FLUO018828-10G
Methyl-2-chlor-4-nitrobenzoat
Methyl-2-chlor-4-nitrobenzoat
Formel:
C₈H₆ClNO₄ Molecular Weight: 215,59 g/mol |
MDL:
MFCD00017013 CAS-Nummer: 13324-11-3 EINECS: 236-362-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...