Methyl-2,4-dimethoxybenzoat

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dimethoxybenzoesäuremethylester

018831-10G
FLUO018831-10GEA 243 EUR
FLUO018831-10G
Methyl-2,4-dimethoxybenzoat
Methyl-2,4-dimethoxybenzoat
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
MDL: MFCD00070649
CAS-Nummer: 2150-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR