Methyl-2,5-dimethoxybenzoat

Lieferant: FLUOROCHEM

Synonyme: 2,5-Dimethoxybenzoesäuremethylester

018832-5G
FLUO018832-5GEA 299 EUR
FLUO018832-5G
Methyl-2,5-dimethoxybenzoat
Methyl-2,5-dimethoxybenzoat
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
MDL: MFCD00060624
CAS-Nummer: 2150-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR