3,4-(Methylendioxy)phenylessigsäure

Lieferant: FLUOROCHEM

Synonyme: (1,3-Benzodioxol-5-yl)acetic acid

018839-5G
FLUO018839-5GEA 42.9 EUR
FLUO018839-5G
3,4-(Methylendioxy)phenylessigsäure
3,4-(Methylendioxy)phenylessigsäure
Formel: C₉H₈O₄
Molecular Weight: 180.16 g/mol
MDL: MFCD00014576
CAS-Nummer: 2861-28-1
EINECS: 220-679-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR